3-Fluoro-4-hydroxybenzaldehyde ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
F0725-1G
F0725-5G
TCIAF0725-1GEA
595
NOK
TCIAF0725-1G
TCIAF0725-5G
3-Fluoro-4-hydroxybenzaldehyde ≥98.0% (by GC)
3-Fluoro-4-hydroxybenzaldehyde
Formel:
C₇H₅FO₂ MW: 140,11 g/mol |
MDL nummer:
MFCD00016626 CAS-nummer: 405-05-0 |
Spesifikasjon Testresultater
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 123.0 to 127.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...