L(+)-Glutamic acid hydrochloride ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: (S)-2-Aminopentanedioic acid hydrochloride, 2-Aminoglutaric acid hydrochloride

G0060-25G G0060-500G
TCIAG0060-25GEA 220 NOK
TCIAG0060-25G TCIAG0060-500G
L(+)-Glutamic acid hydrochloride ≥98.0% (by titrimetric analysis)
L(+)-Glutamic acid hydrochloride
Formel: C₅H₁₀ClNO₄
MW: 183,59 g/mol
MDL nummer: MFCD00012619
CAS-nummer: 138-15-8
EINECS: 205-315-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06a94efa2bdeb37b9b51bc83204c8aa27f4cda67&vid=06a94efa2bdeb37b9b51bc83204c8aa27f4cda67&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +22.0 to +25.0 deg (C=6, H2O)
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR