1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride ≥98.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: 1,3-Dimesityl-1H-imidazol-3-ium chloride, 1,3-Dimesitylimidazolium chloride

D3446-1G D3446-5G
TCIAD3446-1GEA 595 NOK
TCIAD3446-1G TCIAD3446-5G
1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride ≥98.0% (by HPLC)
1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride
Formel: C₂₁H₂₅ClN₂
MW: 340,89 g/mol
Smeltepunkt: 352°C
MDL nummer: MFCD02684541
CAS-nummer: 141556-45-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceea41357e9682763751e9db2e406eed713b43c3&vid=ceea41357e9682763751e9db2e406eed713b43c3&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR