(±)-α-Bromophenylacetic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Danger
|
Synonymer:
α-Bromophenylacetic Acid, DL-α-Bromophenylacetic acid
B0984-25G
B0984-5G
TCIAB0984-25GEA
1375
NOK
TCIAB0984-25G
TCIAB0984-5G
(±)-α-Bromophenylacetic acid ≥98.0% (by GC, titration analysis)
(±)-α-Bromophenylacetic acid
Formel:
C₈H₇BrO₂ MW: 215,0440 g/mol |
MDL nummer:
MFCD00004206 CAS-nummer: 4870-65-9 EINECS: 225-477-4 FN: 3261 ADR: 8,II |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 80.0 to 84.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...