Tetramethyl orthocarbonate ≥97.0% (by GC)

Leverandør: TCI
Danger

Synonymer: Tetramethylorthocarbonate

T1045-25G T1045-5G
TCIAT1045-25GEA 10525 NOK
TCIAT1045-25G TCIAT1045-5G
Tetramethyl orthocarbonate ≥97.0% (by GC)
Tetramethyl orthocarbonate
Formel: C₅H₁₂O₄
MW: 136,15 g/mol
Kokepunkt: 114°C
Flash punkt: 22°C
MDL nummer: MFCD00008473
CAS-nummer: 1850-14-2
EINECS: 217-438-5
FN: 3272
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dcc849bed61766b8ec4c79700a15b48b6fc0404&vid=5dcc849bed61766b8ec4c79700a15b48b6fc0404&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR