2-Aminoethyl dihydrogen phosphate ≥98.0% (by total nitrogen and titration analysis)

Leverandør: TCI
Danger

Synonymer: O-Phosphorylethanolamine

P0279-100G P0279-25G
TCIAP0279-100GEA 1350 NOK
TCIAP0279-100G TCIAP0279-25G
2-Aminoethyl dihydrogen phosphate ≥98.0% (by total nitrogen and titration analysis)
2-Aminoethyl dihydrogen phosphate
Formel: C₂H₈NO₄P
MW: 141,0630 g/mol
Smeltepunkt: 235°C
MDL nummer: MFCD00008178
CAS-nummer: 1071-23-4
EINECS: 213-988-5
FN: 1759
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=278325b8543e783f0e74b5b4eff0898b6b98b753&vid=278325b8543e783f0e74b5b4eff0898b6b98b753&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR