5-Chloro-2-methoxypyridine ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
C2522-1G
C2522-5G
TCIAC2522-1GEA
285
NOK
TCIAC2522-1G
TCIAC2522-5G
5-Chloro-2-methoxypyridine ≥98.0% (by GC)
5-Chloro-2-methoxypyridine
Formel:
C₆H₆ClNO MW: 143,5700 g/mol Kokepunkt: 180°C Flash punkt: 53°C |
MDL nummer:
MFCD06254388 CAS-nummer: 13473-01-3 FN: 1993 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...