Azacyclonol ≥99.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: alpha,alpha-Diphenyl-4-piperidinemethanol, alpha-(4-Piperidyl)benzhydrol, Diphenyl(4-piperidinyl)methanol, α,α-Diphenyl-4-piperidinomethanol, Diphenyl-4-piperidinemethanol

P1369-25G P1369-5G
TCIAP1369-25GEA 1350 NOK
TCIAP1369-25G TCIAP1369-5G
Azacyclonol ≥99.0% (by GC, titration analysis)
Azacyclonol
Formel: C₁₈H₂₁NO
MW: 267,37 g/mol
MDL nummer: MFCD00066980
CAS-nummer: 115-46-8
EINECS: 204-092-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18f9d3078675a1c04703afa5cf2085a864263ea1&vid=18f9d3078675a1c04703afa5cf2085a864263ea1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 161.0 to 164.0 deg-C
Solubility in hot Methanol within almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR