4-Chloro-2,6-dimethoxypyrimidine ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 6-Chloro-2,4-dimethoxypyrimidine

D1385-1G D1385-5G
TCIAD1385-1GEA 325 NOK
TCIAD1385-1G TCIAD1385-5G
4-Chloro-2,6-dimethoxypyrimidine ≥98.0% (by GC)
4-Chloro-2,6-dimethoxypyrimidine
Formel: C₆H₇ClN₂O₂
MW: 174,59 g/mol
MDL nummer: MFCD00006068
CAS-nummer: 6320-15-6
EINECS: 228-669-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 76.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR