2-Amino-4,4,4-trifluorobutyric acid hydrochloride ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: (±)-2-Amino-4,4,4-trifluorobutyric acid hydrochloride

A1367-1G A1367-100MG
TCIAA1367-1GEA 3100 NOK
TCIAA1367-1G TCIAA1367-100MG
2-Amino-4,4,4-trifluorobutyric acid hydrochloride ≥98.0% (by titrimetric analysis)
(±)-2-Amino-4,4,4-trifluorobutyric acid hydrochloride
Formel: C₄H₇ClF₃NO₂
MDL nummer: MFCD00671488
CAS-nummer: 15959-93-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb4533662630feefb80ef22b3700fad3ebe27c5a&vid=d8cb910cf32c74c23ae40911b68218022a797e25&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystaline
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR