2-Ethoxy-3-methylpyrazine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
3-Ethoxy-2-methylpyrazine
M0809-25G
M0809-5G
TCIAM0809-25GEA
2375
NOK
TCIAM0809-25G
TCIAM0809-5G
2-Ethoxy-3-methylpyrazine ≥98.0% (by GC, titration analysis)
2-Ethoxy-3-methylpyrazine
Formel:
C₇H₁₀N₂O MW: 138,167 g/mol Kokepunkt: 88°C Flash punkt: 66°C |
MDL nummer:
MFCD00038025 CAS-nummer: 32737-14-7 EINECS: 251-184-6 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...