6-tert-Butyl-2,4-xylenol ≥97.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 2-(tert-Butyl)-4,6-dimethylphenol

B0903-25G B0903-500G B0903-100G
TCIAB0903-25GEA 285 NOK
TCIAB0903-25G TCIAB0903-500G TCIAB0903-100G
6-tert-Butyl-2,4-xylenol ≥97.0% (by GC)
6-tert-Butyl-2,4-xylenol
Formel: C₁₂H₁₈O
MW: 178,28 g/mol
Kokepunkt: 249°C
MDL nummer: MFCD00002234
CAS-nummer: 1879-09-0
EINECS: 217-533-1
FN: 2811
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94b9f4c00bb61414a236bc7a3d0e378b1a4832fd&vid=94b9f4c00bb61414a236bc7a3d0e378b1a4832fd&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %
Freezing point 20.0 to 24.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR