Tetrabutylammonium azide ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Danger

T0920-25G T0920-5G
TCIAT0920-25GEA 3300 NOK
TCIAT0920-25G TCIAT0920-5G
Tetrabutylammonium azide ≥95.0% (by titrimetric analysis)
Tetrabutylammonium azide
Formel: C₁₆H₃₆N₄
Smeltepunkt: 85°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00060069
CAS-nummer: 993-22-6
FN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53f7d23ec3f90efbd42943c1511666d8ec0e984e&vid=53f7d23ec3f90efbd42943c1511666d8ec0e984e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR