2-Mercaptopyridine-N-oxide zinc salt ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: Bis(2-pyridylthio) Zinc 1,1'-Dioxide, Zinc Pyrithione, 2-Pyridinethiol 1-Oxide Zinc salt, 1-Hydroxypyridine-2-thione Zinc salt

M0633-25G M0633-500G M0633-100G
TCIAM0633-25GEA 635 NOK
TCIAM0633-25G TCIAM0633-500G TCIAM0633-100G
2-Mercaptopyridine-N-oxide zinc salt ≥98.0% (by titrimetric analysis)
2-Mercaptopyridine-N-oxide zinc salt
Formel: C₁₀H₈N₂O₂S₂Zn
MW: 317,72 g/mol
MDL nummer: MFCD00067336
CAS-nummer: 13463-41-7
EINECS: 236-671-3
FN: 2811
ADR: 6.1,II
Merck Indeks: 14,07994
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=708528199b9eec3a9d1113ce3213adde6a36be1e&vid=708528199b9eec3a9d1113ce3213adde6a36be1e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Solubility in Pyridine almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR