4-Bromo-N,N-diethylaniline ≥97.0% (by GC)
Leverandør: TCI
|
Warning
|
B3027-25G
B3027-5G
TCIAB3027-25GEA
2175
NOK
TCIAB3027-25G
TCIAB3027-5G
4-Bromo-N,N-diethylaniline ≥97.0% (by GC)
4-Bromo-N,N-diethylaniline
Formel:
C₁₀H₁₄BrN MW: 228,13 g/mol Kokepunkt: 215°C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00013530 CAS-nummer: 2052-06-4 EINECS: 218-140-8 ADR: 6.1,III |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to lump |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...