(R)-(+)-β-Methylphenethylamine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Danger
|
Synonymer:
(R)-(+)-beta-Methylphenethylamine, (R)-(+)-2-Phenyl-1-propylamine
M1746-25G
M1746-5G
TCIAM1746-25GEA
3000
NOK
TCIAM1746-25G
TCIAM1746-5G
(R)-(+)-β-Methylphenethylamine ≥98.0% (by GC, titration analysis)
(R)-(+)-β-Methylphenethylamine
Formel:
C₉H₁₃N Kokepunkt: 102°C Flash punkt: 81°C |
MDL nummer:
MFCD00216740 CAS-nummer: 28163-64-6 FN: 2735 ADR: 8,II |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Optical purity (LC) | min. 80.0 ee% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | +30.0 to +35.0 deg (C=1, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...