N-(Phenylthio)phthalimide ≥98.0% (by GC, total nitrogen)

Leverandør: TCI

Synonymer: 2-(Phenylthio)isoindoline-1,3-dione

P1254-25G P1254-5G
TCIAP1254-25GEA 4450 NOK
TCIAP1254-25G TCIAP1254-5G
N-(Phenylthio)phthalimide ≥98.0% (by GC, total nitrogen)
N-(Phenylthio)phthalimide
Formel: C₁₄H₉NO₂S
MW: 255,2918 g/mol
MDL nummer: MFCD00192396
CAS-nummer: 14204-27-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed7d902d44f35408b41856714cfea25fdda9de1e&vid=ed7d902d44f35408b41856714cfea25fdda9de1e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 161.0 to 166.0 deg-C
Solubility in THF almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR