Methyl-6-bromo-2-naphthoate ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
Methyl 6-bromo-2-naphthoate
B2202-25G
B2202-5G
TCIAB2202-25GEA
2325
NOK
TCIAB2202-25G
TCIAB2202-5G
Methyl-6-bromo-2-naphthoate ≥98.0% (by GC)
Methyl-6-bromo-2-naphthoate
Formel:
C₁₂H₉BrO₂ MW: 265,11 g/mol |
MDL nummer:
MFCD00100408 CAS-nummer: 33626-98-1 |
Spesifikasjon Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 123.0 to 126.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...