2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid) ≥98%
Leverandør: Thermo Scientific
Warning
Synonymer: 2,2-Dimethyl-1,3-dioxane-4,6-dione
A15603.14 A15603.22 A15603.36
A15603.14EA 700 NOK
A15603.14 A15603.22 A15603.36
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid) ≥98%
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid)
Purity: 98% CAS No.: 2033-24-1 Molecular Formula: C6H8O4 Formula Weight: 144.13 Melting Point: 92-96° Beilstein Reference: 117310 Merck Reference: 14,5817 MDL No.: MFCD00006638
Formel: C₆H₈O₄
MW: 144,13 g/mol
Smeltepunkt: 92…96 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006638
CAS-nummer: 2033-24-1
EINECS: 217-992-8
Merck Indeks: 14,5817