HPLC columns, Acclaim™ mixed-mode HILIC-1
Leverandør: Thermo Fisher Scientific
066843 066844 066847 069694 069706 070090 070091 071912 071913
554-4455EA 12575 NOK
554-4455 554-4456 554-4457 555-7128 555-7138 554-4460 554-4461 554-4462 555-7170
HPLC columns, Acclaim™ mixed-mode HILIC-1
Kromatografikolonner
These columns utilise a unique silica-based, mixed-mode stationary phase that combines both reversed phase (RP) and hydrophilic interaction liquid chromatography (HILIC) properties. The column's functional group features a hydrophobic alkyl chain and hydrophilic diol functionality. This combination allows both hydrophobic interaction and hydrophilic interaction to be utilised to optimise separations.
  • Retains highly polar molecules
  • Unique selectivity complementary to RP columns
  • Superior performance compared to conventional diol-based columns
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Maks. trykk 8500 psi (4800 psi for 3,0×50 mm column)
pH område
Temperaturgrense