Solid phase extraction cartridges, EnvirElut™
Leverandør: VARIAN
12272001 12272020 12272005 12272004 12272011 12102158
VARI12272001EA 9225 NOK
VARI12272001 VARI12272020 VARI12272005 VARI12272004 VARI12272011 VARI12102158
Solid phase extraction cartridges, EnvirElut™
Kromatografikolonner
EnvirElut™ sorbents are specially designed for the extraction of a wide range of compounds from aqueous matrices. EnvirElut™ PAH and Pesticides are available in standard SPE straight barrel cartridges, which can be used on conventional vacuum manifolds such as the Vac Elut SPS 24.
  • Extreme purity offers cleanliness in extract
  • High capacity allows for the processing of large sample volumes
  • Broad compound specificity
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Karbonbelastning 18,0%
Kappet ved endene Nei
Overflate areal 500 m²/g