2,6-Difluorophenylboronic acid (contains varying amounts of Anhydride)
Leverandør: TCI
|
Warning
|
Synonymer:
Dihydroxy(2,6-difluorophenyl)borane, 2,6-Difluorobenzeneboronic acid, (2,6-Difluorophenyl)metaboric acid
D3087-1G
D3087-5G
D3087-25G
TCIAD3087-1GEA
220
NOK
TCIAD3087-1G
TCIAD3087-5G
TCIAD3087-25G
2,6-Difluorophenylboronic acid (contains varying amounts of Anhydride)
2,6-Difluorophenylboronic acid
Formel:
C₆H₅BF₂O₂ MW: 157,91 g/mol |
MDL nummer:
MFCD00792436 CAS-nummer: 162101-25-9 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 90.0 to 113.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...