6-Aminokapronsyre ≥98.0% (by titrimetric analysis) for biochemical research

Leverandør: TCI
Warning

Synonymer: 6-Aminocaproic Acid, H-Eahx-OH, ε-Amino-n-caproic acid, 6-Aminohexanoic acid, Aminocaproic Acid

A2255-25G A2255-5G
TCIAA2255-25GEA 350 NOK
TCIAA2255-25G TCIAA2255-5G
6-Aminokapronsyre ≥98.0% (by titrimetric analysis) for biochemical research
6-Aminokapronsyre
Formel: C₆H₁₃NO₂
MW: 131,17 g/mol
Kokepunkt: 257 °C (1013 hPa)
Smeltepunkt: 204°C
Flash punkt: 207 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00008238
CAS-nummer: 60-32-2
EINECS: 200-469-3
Merck Indeks: 14,00432
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2a793eb45f7423924f34cd9682b528ab8bfa6e6&vid=63ddedc2e1806ac8ee8350ff5a503d5eced62a3c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency
Blotting test to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR