2-Iodopyridine ≥98.0% (by GC) stabilised
Leverandør: TCI
|
Warning
|
Stabilisering:
Stabilised with Na₂S₂O₃
I0533-25G
I0533-5G
TCIAI0533-25GEA
2800
NOK
TCIAI0533-25G
TCIAI0533-5G
2-Iodopyridine ≥98.0% (by GC) stabilised
2-Iodopyridine
Formel:
C₅H₄IN MW: 205 g/mol Kokepunkt: 102°C Smeltepunkt: 118…120 °C Tetthet: 1,928 g/cm³ (25 °C) |
MDL nummer:
MFCD00464928 CAS-nummer: 5029-67-4 |
Spesifikasjon Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...