1-Pentansulfonsyre natriumsalt ≥98.0% (by titrimetric analysis) for ioneparkromatografi
Leverandør: TCI
Synonymer: Sodium 1-pentanesulphonate, Sodium pentane-1-sulphonate, Pentane-1-sulphonic acid sodium salt, Sodium pentanesulphonate, 98%, Sodium pentanesulphonate
I0343-100G I0343-25G I0343-5G
TCIAI0343-100GEA 2700 NOK
TCIAI0343-100G TCIAI0343-25G TCIAI0343-5G
1-Pentansulfonsyre natriumsalt ≥98.0% (by titrimetric analysis) for ioneparkromatografi
1-Pentansulfonsyre natriumsalt
Formel: C₅H₁₁O₃SNa
MW: 174,2 g/mol
Smeltepunkt: > 300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007541
CAS-nummer: 22767-49-3
EINECS: 245-208-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66d840d652ad09ac81b2a6396c594b1c259e2c79&vid=dca2905e8eaa2531e8dfd202d2b1ca647ea34032&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Absorbance (210nm) max. 1.000
Absorbance (230nm) max. 0.300
Absorbance (254nm) max. 0.130