Pyrazine-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: Pyrazinecarboxylic acid, 2-Pyrazinecarboxylic Acid

P0940-25G P0940-500G
TCIAP0940-25GEA 350 NOK
TCIAP0940-25G TCIAP0940-500G
Pyrazine-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Pyrazine-2-carboxylic acid
Formel: C₅H₄N₂O₂
MW: 124,1 g/mol
Kokepunkt: 313 °C (1013 hPa)
MDL nummer: MFCD00006130
CAS-nummer: 98-97-5
EINECS: 202-718-1
Merck Indeks: 14,08970
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77e42f53f1e964a259ba4ac0d75f8f0d5a4292c8&vid=77e42f53f1e964a259ba4ac0d75f8f0d5a4292c8&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR