Pentyl-4-hydroxybenzoate ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
Synonymer: Pentylparaben, 4-Hydroxybenzoic acid Amyl Ester, 4-Hydroxybenzoic acid pentyl ester, Pentyl parahydroxybenzoate, Pentyl 4-Hydroxybenzoate, 4-Hydroxybenzoic acid Pentyl Ester, Amyl 4-Hydroxybenzoate, Amylparaben, n-Pentyl 4-hydroxybenzoate
H0208-10G H0208-25G
TCIAH0208-10GEA 595 NOK
TCIAH0208-10G TCIAH0208-25G
Pentyl-4-hydroxybenzoate ≥98.0% (by GC, titration analysis)
Pentyl 4-hydroxybenzoate
Formel: C₁₂H₁₆O₃
MW: 208,26 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00016479
CAS-nummer: 6521-29-5
EINECS: 229-408-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=169ebc8b5b1412aab1523d32d47453b732b136e7&vid=169ebc8b5b1412aab1523d32d47453b732b136e7&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency