2-(2-Hydroxyethyl)pyridine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
Synonymer: 2-Pyridineethanol, 2-(2-Pyridyl)ethanol
H0246-25ML H0246-500ML
TCIAH0246-25MLEA 340 NOK
TCIAH0246-25ML TCIAH0246-500ML
2-(2-Hydroxyethyl)pyridine ≥98.0% (by GC, titration analysis)
2-(2-Hydroxyethyl)pyridine
Formel: C₇H₉NO
MW: 123,15 g/mol
Kokepunkt: 139°C
Smeltepunkt: –7,8 °C
Tetthet: 1,1111 g/cm³ (4 °C)
Flash punkt: 92°C
MDL nummer: MFCD00006364
CAS-nummer: 103-74-2
EINECS: 203-140-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e6672ba99581ff5d7c688da22d71ec096020c8b&vid=6e6672ba99581ff5d7c688da22d71ec096020c8b&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %