α-Methylstyrene ≥99.0% (by GC) stabilised

Leverandør: TCI
Warning

Synonymer: 2-Phenylpropene, Isopropenylbenzene

Stabilisering: Stabilised with 4-tert-butylpyrocatechol (TBC)

M0429-25ML M0429-500ML
TCIAM0429-25MLEA 220 NOK
TCIAM0429-25ML TCIAM0429-500ML
α-Methylstyrene ≥99.0% (by GC) stabilised
α-Methylstyrene
Formel: C₉H₁₀
MW: 118.18 g/mol
Kokepunkt: 162°C
Smeltepunkt: –23°C
Flash punkt: 48°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00008859
CAS-nummer: 98-83-9
EINECS: 202-705-0
FN: 2303
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1131e388cd00ec667f273b1d197097668c5e2c57&vid=c6b1e659c82c7ddd7de5e33f20301c75db3d1fab&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR