Nicotinaldehyde ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Pyridine-3-carboxaldehyde, 3-Pyridinecarboxaldehyde, 3-Pyridinecarbaldehyde
N0090-25ML
N0090-100ML
TCIAN0090-25MLEA
700
NOK
TCIAN0090-25ML
TCIAN0090-100ML
Nicotinaldehyde ≥98.0% (by GC)
Nicotinaldehyde
Formel:
C₆H₅NO MW: 107,11 g/mol Kokepunkt: 97°C Flash punkt: 43°C |
MDL nummer:
MFCD00006382 CAS-nummer: 500-22-1 EINECS: 207-900-4 FN: 1989 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | Colorless to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...