N-Methyl-1,3-propanediamine ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: N-Methylpropane-1,3-diamine, 3-Methylamino propylamine

M0599-25ML M0599-500ML M0599-100ML
TCIAM0599-25MLEA 245 NOK
TCIAM0599-25ML TCIAM0599-500ML TCIAM0599-100ML
N-Methyl-1,3-propanediamine ≥98.0% (by GC)
N-Methyl-1,3-propanediamine
Formel: C₄H₁₂N₂
MW: 88,15 g/mol
Kokepunkt: 143°C
Flash punkt: 36°C
MDL nummer: MFCD00008209
CAS-nummer: 6291-84-5
EINECS: 228-544-6
FN: 2734
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29f1602ff1522a93607350bc8b637695d5f1077c&vid=29f1602ff1522a93607350bc8b637695d5f1077c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR