cis-4-tert-Butylcyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

B2631-25G B2631-5G
TCIAB2631-25GEA 6475 NOK
TCIAB2631-25G TCIAB2631-5G
cis-4-tert-Butylcyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
cis-4-tert-Butylcyclohexanecarboxylic acid
Formel: C₁₁H₂₀O₂
CAS-nummer: 943-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e168c14ac45de3709a3c50251d27380db33ac310&vid=e168c14ac45de3709a3c50251d27380db33ac310&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 116.0 to 121.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR