(2R)-(-)-2,3-Epoxypropyl toluene-4-sulphonate ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
(2R)-(-)-Glycidyl p-toluenesulphonate, (2R)-(-)-Glycidyl tosylate
T1611-25G
T1611-5G
TCIAT1611-25GEA
1200
NOK
TCIAT1611-25G
TCIAT1611-5G
(2R)-(-)-2,3-Epoxypropyl toluene-4-sulphonate ≥98.0% (by GC)
(2R)-(-)-2,3-Epoxypropyl toluene-4-sulphonate
Formel:
C₁₀H₁₂O₄S MW: 228,2649 g/mol |
MDL nummer:
MFCD00010834 CAS-nummer: 113826-06-5 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Optical purity (LC) | min. 95.0 ee% |
Melting point | 47.0 to 50.0 deg-C |
Specific rotation [a]20/D | -18.0 to -20.0 deg (C=2.5, CH3CN) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...