1-(2-Hydroxyethyl)pyrrole ≥99.0% (by GC)

Leverandør: TCI
Warning

H0826-1G H0826-5G
TCIAH0826-1GEA 1150 NOK
TCIAH0826-1G TCIAH0826-5G
1-(2-Hydroxyethyl)pyrrole ≥99.0% (by GC)
1-(2-Hydroxyethyl)pyrrole
Formel: C₆H₉NO
Kokepunkt: 215°C
MDL nummer: MFCD00191451
CAS-nummer: 6719-02-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36c081097c2ff8f87a8c62d286774bb08c701449&vid=36c081097c2ff8f87a8c62d286774bb08c701449&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR