Flavanone ≥98%
Leverandør: Thermo Scientific
Synonymer:
2-Phenylchroman-4-one
B22587.09
B22587.18
B22587.09EA
675
NOK
B22587.09
B22587.18
Flavanone ≥98%
Flavanone
Purity: 98+%
CAS No.: 487-26-3
Molecular Formula: C15H12O2
Formula Weight: 224.26
Melting Point: 75-79°
Beilstein Reference: 183227
Formel:
C₁₅H₁₂O₂ MW: 224,26 g/mol Smeltepunkt: 75…79 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00006841 CAS-nummer: 487-26-3 EINECS: 207-654-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...