6-Methoxyflavanone ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
6-Methoxy-2-phenylchroman-4-one
M1403-1G
M1403-5G
TCIAM1403-1GEA
500
NOK
TCIAM1403-1G
TCIAM1403-5G
6-Methoxyflavanone ≥98.0% (by GC)
6-Methoxyflavanone
Formel:
C₁₆H₁₄O₃ MW: 254.29 g/mol |
MDL nummer:
MFCD00017484 CAS-nummer: 3034-04-6 |
|
Spesifikasjon Testresultater
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 141.0 to 145.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...