6-Hydroxyflavanone ≥98%

Leverandør: Thermo Scientific
Warning

Synonymer: 6-Hydroxy-2-phenylchroman-4-one

B23755.03 B23755.06
B23755.03EA 650 NOK
B23755.03 B23755.06
6-Hydroxyflavanone ≥98%
6-Hydroxyflavanone

Purity: 98+% CAS No.: 4250-77-5 Molecular Formula: C15H12O3 Formula Weight: 240.25 Melting Point: ca 220° dec. Beilstein Reference: 87354

Formel: C₁₅H₁₂O₃
MW: 240,25 g/mol
Smeltepunkt: ∼ 220 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00017485
CAS-nummer: 4250-77-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR