Laboratory jacks
Leverandør: BOCHEM
11120 11130 11140 11115 11180 11190 11132 11122 11123 11133 11143
245-1191EA 4800 NOK
245-1191 245-1215 245-1192 245-1190 245-1193 245-1194 245-0028 245-0027 BOCH11123 BOCH11133 BOCH11143
Laboratory jacks
Laboratoriejekker
These lab jacks feature an improved scissor technology with reduced tolerances. The scissor construction is made of ¹⁸/₁₀ stainless steel.
  • Ergonomic round knob (either made of plastic or metal)
  • Smaller models up to 240×240 mm have a simple adjusting wheel which can be operated by hand
  • Larger models from 300×300 mm on should be used with a ratchet, which connects with the special adjusting wheel (ratchet is included)
  • Models with metal knob are autoclavable
DIN 12897
Sertifiseringer: DIN 12897