3-(Propylthio)propene ≥97.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Allyl Propyl sulphide
A1061-25ML
A1061-5ML
TCIAA1061-25MLEA
3675
NOK
TCIAA1061-25ML
TCIAA1061-5ML
3-(Propylthio)propene ≥97.0% (by GC)
3-(Propylthio)propene
Formel:
C₆H₁₂S MW: 116,23 g/mol Kokepunkt: 140°C Tetthet: 0,87 g/cm³ (20 °C) |
MDL nummer:
MFCD00015220 CAS-nummer: 27817-67-0 EINECS: 248-673-1 FN: 1993 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...