Piperazin, vannfri ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Danger
P0446-25G P0446-500G
TCIAP0446-25GEA 300 NOK
TCIAP0446-25G TCIAP0446-500G
Piperazin, vannfri ≥98.0% (by GC, titration analysis)
Piperazin
Formel: C₄H₁₀N₂
MW: 86,14 g/mol
Kokepunkt: 146 °C (1013 hPa)
Smeltepunkt: 44…46 °C
Tetthet: 1,1 g/cm³ (20 °C)
Flash punkt: 65 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005953
CAS-nummer: 110-85-0
EINECS: 203-808-3
FN: 2579
ADR: 8,III
Merck Indeks: 14,07464
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7121c109cda0e28fd9eee6f3fdee46138495a2b&vid=50e812a0b33534f9769755db89d93fca83150945&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Water almost transparency