3-Fluorophenylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Warning

Synonymer: 3-Fluorobenzeneboronic acid, (3-Fluorophenyl)boronic acid, Dihydroxy(3-fluorophenyl)borane, (3-Fluorophenyl)metaboric acid

F0404-25G F0404-5G
TCIAF0404-25GEA 745 NOK
TCIAF0404-25G TCIAF0404-5G
3-Fluorophenylboronic acid (contains varying amounts of Anhydride)
3-Fluorophenylboronic acid
Formel: C₆H₆BFO₂
MW: 139,92 g/mol
Smeltepunkt: 216…218 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00236042
CAS-nummer: 768-35-4
EINECS: 476-720-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96c3d517cd33a6a822102d3f0475411b279c4380&vid=96c3d517cd33a6a822102d3f0475411b279c4380&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) 95.0 to 115.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR