4-Ethoxyphenylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Warning

Synonymer: 4-Ethoxybenzeneboronic acid, (4-Ethoxyphenyl)metaboric acid, Dihydroxy(4-ethoxyphenyl)borane

E0725-5G E0725-1G
TCIAE0725-5GEA 700 NOK
TCIAE0725-5G TCIAE0725-1G
4-Ethoxyphenylboronic acid (contains varying amounts of Anhydride)
4-Ethoxyphenylboronic acid
Formel: C₈H₁₁BO₃
MW: 165,98 g/mol
MDL nummer: MFCD00674028
CAS-nummer: 22237-13-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03303d5ecc2d8c4b3c6368fdefd8356e3992f5f4&vid=03303d5ecc2d8c4b3c6368fdefd8356e3992f5f4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR