Etyl (S)-(-)-laktat ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: Ethyl (S)-(-)-2-hydroxypropionate, Ethyl L(-)-lactate, (S)-Ethyl 2-hydroxypropanoate, (S)-(-)-Ethyl lactate

L0162-25G L0162-500G
TCIAL0162-25GEA 220 NOK
TCIAL0162-25G TCIAL0162-500G
Etyl (S)-(-)-laktat ≥98.0% (by GC)
Etyl (S)-(-)-laktat
Formel: C₅H₁₀O₃
MW: 118,13 g/mol
Kokepunkt: 70°C
Smeltepunkt: –25 °C
Tetthet: 1,042 g/cm³ (20 °C)
Flash punkt: 46°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004518
CAS-nummer: 687-47-8
EINECS: 211-694-1
FN: 1192
ADR: 3,III
Merck Indeks: 12,03850
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8c06106b0bd144ee7b07bc2391cc42b95e27970&vid=d8c06106b0bd144ee7b07bc2391cc42b95e27970&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D -10.0 to -11.5 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR