Methacrylic acid ≥99.0% (by GC) stabilised

Leverandør: TCI
Danger

Synonymer: 2-Methylpropenoic acid

Stabilisering: Stabilised with 4-methoxyphenol (MEHQ)

M0079-500G M0079-25G
TCIAM0079-500GEA 435 NOK
TCIAM0079-500G TCIAM0079-25G
Methacrylic acid ≥99.0% (by GC) stabilised
Methacrylic acid
Formel: C₄H₆O₂
MW: 86,09 g/mol
Kokepunkt: 160°C
Smeltepunkt: 15 °C
Tetthet: 1,02 g/cm³ (20 °C)
Flash punkt: 77°C
MDL nummer: MFCD00002651
CAS-nummer: 79-41-4
EINECS: 201-204-4
FN: 2531
ADR: 8,II
Merck Indeks: 14,05941
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3994515d4a04cc85f59e28fcaa566d3032bd5a42&vid=3994515d4a04cc85f59e28fcaa566d3032bd5a42&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR