3-Tolylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Warning

Synonymer: (3-Methylphenyl)metaboric acid, Dihydroxy(3-methylphenyl)borane, 3-Methylbenzeneboronic acid, 3-Methylphenylboronic acid, m-Tolylboronic acid

M1314-1G M1314-25G M1314-5G
TCIAM1314-1GEA 220 NOK
TCIAM1314-1G TCIAM1314-25G TCIAM1314-5G
3-Tolylboronic acid (contains varying amounts of Anhydride)
3-Tolylboronic acid
Formel: C₇H₉BO₂
MW: 135,96 g/mol
Smeltepunkt: 165 °C
MDL nummer: MFCD00040198
CAS-nummer: 17933-03-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12296523c6c19989b00dd1755e564c62154b0df6&vid=12296523c6c19989b00dd1755e564c62154b0df6&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) 97.0 to 115.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR