(±)-γ-Undecalactone ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: (±)-Undecan-4-olide, Gamma-undecalactone, Undecanoic gamma-Lactone

U0003-25ML U0003-500ML
TCIAU0003-25MLEA 220 NOK
TCIAU0003-25ML TCIAU0003-500ML
(±)-γ-Undecalactone ≥98.0% (by GC)
(±)-γ-Undecalactone
Formel: C₁₁H₂₀O₂
MW: 184,28 g/mol
Kokepunkt: 110°C
Tetthet: 0,9414 g/cm³ (20 °C)
Flash punkt: 156°C
MDL nummer: MFCD00005405
CAS-nummer: 104-67-6
EINECS: 203-225-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca9d2565175ac1412dfc086b21863e30bcae68f5&vid=ca9d2565175ac1412dfc086b21863e30bcae68f5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR