2-Chloro-4-pyridineboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Warning

Synonymer: (2-Chloropyridin-4-yl)boronic acid, 2-Chloro-4-pyridinylboronic acid, 2-Chloropyridine-4-boronic acid

C2589-1G C2589-5G
TCIAC2589-1GEA 525 NOK
TCIAC2589-1G TCIAC2589-5G
2-Chloro-4-pyridineboronic acid (contains varying amounts of Anhydride)
2-Chloro-4-pyridineboronic acid
Formel: C₅H₅BClNO₂
MW: 157,36 g/mol
Smeltepunkt: 210°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD03788416
CAS-nummer: 458532-96-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88a70b8cc2e4f44b84818c882a551fd693da5a39&vid=88a70b8cc2e4f44b84818c882a551fd693da5a39&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 113.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR