1,1,1-Trifluoracetylaceton ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1,1,1-Trifluoropentane-2,4-dione, 1,1,1-Trifluoro-2,4-pentanedione

T0434-100G T0434-25G
TCIAT0434-100GEA 2600 NOK
TCIAT0434-100G TCIAT0434-25G
1,1,1-Trifluoracetylaceton ≥98.0% (by GC)
1,1,1-Trifluoracetylaceton
Formel: C₅H₅F₃O₂
MW: 154,09 g/mol
Kokepunkt: 101°C
Tetthet: 1,28 g/cm³ (20 °C)
Flash punkt: 26°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00000427
CAS-nummer: 367-57-7
EINECS: 206-698-5
FN: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=115ace5a8d5a94df35b603679a20fcc8daef0060&vid=546d01c6fbc823213db03b8c9fd1ebb4e7c8dcd3&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR