4-Methyl-2-pentenoic acid ≥97.0% (by GC, titration analysis) stabilised

Leverandør: TCI
Danger


Stabilisering: stabilisert med hydroquinon

M0268-5ML M0268-25ML
TCIAM0268-5MLEA 730 NOK
TCIAM0268-5ML TCIAM0268-25ML
4-Methyl-2-pentenoic acid ≥97.0% (by GC, titration analysis) stabilised
4-Methyl-2-pentenoic acid
Formel: C₆H₁₀O₂
MW: 114,14 g/mol
Kokepunkt: 212°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00043804
CAS-nummer: 10321-71-8
EINECS: 233-706-4
FN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be5beac5890ecedb80db9ad3777b7507f626745d&vid=be5beac5890ecedb80db9ad3777b7507f626745d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR