2-Bromo-3,3,3-trifluoropropene ≥97.0% (by GC) stabilised

Leverandør: TCI
Warning

Synonymer: 2-Bromo-3,3,3-trifluoro-1-propene

Stabilisering: Stabilised with copper

B1801-25G B1801-5G
TCIAB1801-25GEA 1775 NOK
TCIAB1801-25G TCIAB1801-5G
2-Bromo-3,3,3-trifluoropropene ≥97.0% (by GC) stabilised
2-Bromo-3,3,3-trifluoropropene
Formel: C₃H₂BrF₃
MW: 174,95 g/mol
Kokepunkt: 34°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00077469
CAS-nummer: 1514-82-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21676d4c23503f733296ee0e94de4f8ae8ebf070&vid=21676d4c23503f733296ee0e94de4f8ae8ebf070&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR